Search
Search

agr2| Gene Symbol: agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016627.3 | NP_001016627.1 | 5 - 6 | 380 | 66 | |
| XM_012964714.2 | XP_012820168.1 | 6 - 7 | 487 | 66 | ||
| GenBank mRNA | CR760462.2 | - | 5 - 6 | 382 | 66 | |
| CR942718.2 | - | 1789 | 66 | |||
Set Membership: |
|