Search
Search

agr2| Gene Symbol: agr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016627.3 | NP_001016627.1 | 6 - 7 | 463 | 73 | |
| XM_012964714.2 | XP_012820168.1 | 7 - 8 | 570 | 73 | ||
| GenBank mRNA | CR760462.2 | - | 6 - 7 | 465 | 73 | |
| CR942718.2 | - | 1872 | 73 | |||
Set Membership: |
|