Search
Search

slc20a2| Gene Symbol: slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016904.2 | NP_001016904.1 | 6 - 7 | 1099 | 69 | |
| XM_012962996.2 | XP_012818450.1 | 6 - 7 | 1127 | 69 | ||
| XM_012963000.2 | XP_012818454.1 | 6 - 7 | 1103 | 69 | ||
| XM_012963008.2 | XP_012818462.1 | 6 - 7 | 1180 | 69 | ||
| GenBank mRNA | CR848513.2 | - | 6 - 7 | 1099 | 69 | |
Set Membership: |
|