Search
Search

olfm4| Gene Symbol: olfm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017043.2 | NP_001017043.1 | 1 - 2 | 59 | 92 | |
| XM_012968462.2 | XP_012823916.1 | 1 - 2 | 111 | 92 | ||
| GenBank mRNA | CR761859.2 | - | 1 - 2 | 59 | 92 | |
Set Membership: |
|