Search
Search

ndufb3| Gene Symbol: ndufb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017212.3 | NP_001017212.1 | 2 - 3 | 255 | 64 | |
| GenBank mRNA | AL659532.2 | - | 2 - 3 | 229 | 64 | |
| BC135381.1 | - | 159 | 64 | |||
| CN083461.1 | - | 2 - 3 | 258 | 64 | ||
| CR760440.2 | - | 2 - 3 | 201 | 64 | ||
| DN010310.2 | - | 1 - 2 | 235 | 64 | ||
Set Membership: |
|