Search
Search

ddx23| Gene Symbol: ddx23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017213.2 | NP_001017213.1 | 2 - 3 | 194 | 101 | |
| XM_012957044.2 | XP_012812498.1 | 17 - 18 | 2386 | 101 | ||
| XM_012957045.2 | XP_012812499.1 | 17 - 18 | 2283 | 101 | ||
| XM_012957047.2 | XP_012812501.1 | 17 - 18 | 2383 | 101 | ||
| GenBank mRNA | BC153691.1 | - | 5 - 6 | 733 | 101 | |
| CR760437.2 | - | 2 - 3 | 194 | 101 | ||
Set Membership: |
|