Search
Search

gdpd1| Gene Symbol: gdpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017323.2 | NP_001017323.1 | 5 - 6 | 504 | 107 | |
| XM_018091285.1 | XP_017946774.1 | 5 - 6 | 1220 | 107 | ||
| XM_018091286.1 | XP_017946775.1 | 5 - 6 | 1219 | 107 | ||
| XM_018091287.1 | XP_017946776.1 | 5 - 6 | 1220 | 107 | ||
| GenBank mRNA | CR848636.2 | - | 5 - 6 | 504 | 107 | |
Set Membership: |
|