Search
Search

ctu2| Gene Symbol: ctu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039740.1 | NP_001034829.1 | 7 - 8 | 797 | 90 | |
| XM_012961021.1 | XP_012816475.1 | 8 - 9 | 923 | 90 | ||
| XM_012961023.1 | XP_012816477.1 | 8 - 9 | 925 | 90 | ||
| XM_018093032.1 | XP_017948521.1 | 8 - 9 | 923 | 90 | ||
| GenBank mRNA | BC089700.1 | - | 7 - 8 | 774 | 90 | |
| BC167302.1 | - | 7 - 8 | 779 | 90 | ||
| BC170700.1 | - | 7 - 8 | 774 | 90 | ||
| BC170702.1 | - | 7 - 8 | 774 | 90 | ||
| CR762362.2 | - | 7 - 8 | 797 | 90 | ||
Set Membership: |
|