Search
Search

sarnp| Gene Symbol: sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016484.3 | NP_001016484.1 | 4 - 5 | 528 | 83 | |
| XM_012956973.1 | XP_012812427.1 | 4 - 5 | 646 | 83 | ||
| GenBank mRNA | BC121712.1 | - | 4 - 5 | 528 | 83 | |
| BC158968.1 | - | 4 - 5 | 541 | 83 | ||
| CR760849.2 | - | 4 - 5 | 246 | 83 | ||
Set Membership: |
|