Search
Search

bmper| Gene Symbol: bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044402.2 | NP_001037867.1 | 4 - 5 | 417 | 100 | |
| XM_012964815.2 | XP_012820269.1 | 5 - 6 | 4819 | 100 | ||
| XM_012964816.2 | XP_012820270.1 | 5 - 6 | 4816 | 100 | ||
| GenBank mRNA | AY768703.1 | - | 4 - 5 | 418 | 100 | |
| BC135872.1 | - | 4 - 5 | 502 | 100 | ||
Set Membership: |
|