Search
Search

mzt2b| Gene Symbol: mzt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045607.1 | NP_001039072.1 | 2 - 3 | 392 | 88 | |
| GenBank mRNA | BC155371.1 | - | 2 - 3 | 401 | 88 | |
| CR855602.2 | - | 2 - 3 | 392 | 88 | ||
Set Membership: |
|