Search
Search

tctn2| Gene Symbol: tctn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045666.1 | NP_001039131.1 | 6 - 7 | 778 | 71 | |
| GenBank mRNA | BC121278.1 | - | 6 - 7 | 800 | 71 | |
| CR942490.2 | - | 6 - 7 | 778 | 71 | ||
Set Membership: |
|