Search
Search

LOC734042| Gene Symbol: LOC734042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045726.1 | NP_001039191.1 | 4 - 5 | 617 | 84 | |
| XM_018091325.1 | XP_017946814.1 | 3 - 4 | 416 | 84 | ||
| GenBank mRNA | CR855665.1 | - | 4 - 5 | 617 | 84 | |
Set Membership: |
|