Search
Search

nme4| Gene Symbol: nme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045774.1 | NP_001039239.1 | 1 - 2 | 111 | 66 | |
| GenBank mRNA | BC158417.1 | - | 1 - 2 | 141 | 66 | |
| CR942753.2 | - | 1 - 2 | 111 | 66 | ||
Set Membership: |
|