Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078695.1 | NP_001072163.1 | 4 - 5 | 900 | 58 | |
GenBank mRNA | BC122981.1 | - | 4 - 5 | 900 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |