Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Xt03713934_g1
          See other ELN1 GE Assays ›
          Gene Symbol:
          eln1
          Gene Name:
          tropoelastin 1
          Gene Aliases:
          eln
          Chromosome Location:
          Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
          Species:
          Xenopus tropicalis
          Species Specific ID (Flybase ID):
          -
          Assay ID Xt03713934_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: eln1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001078709.1 NP_001072177.1 17 - 18 1104 55
          XM_012957183.2 XP_012812637.1 19 - 20 1290 55
          XM_012957184.2 XP_012812638.1 18 - 19 1260 55
          XM_012957185.2 XP_012812639.1 19 - 20 1290 55
          XM_012957188.1 XP_012812642.1 19 - 20 1219 55
          XM_012957189.2 XP_012812643.1 17 - 18 1197 55
          XM_012957190.2 XP_012812644.1 17 - 18 1169 55
          XM_012957191.2 XP_012812645.1 19 - 20 1291 55
          XM_012957192.2 XP_012812646.1 19 - 20 1291 55
          XM_012957193.1 XP_012812647.1 19 - 20 1292 55
          XM_012957194.2 XP_012812648.1 19 - 20 1290 55
          XM_012957195.2 XP_012812649.1 16 - 17 1139 55
          XM_012957196.1 XP_012812650.1 19 - 20 1291 55
          XM_012957197.1 XP_012812651.1 19 - 20 1291 55
          XM_012957199.2 XP_012812653.1 15 - 16 1076 55
          XM_012957200.1 XP_012812654.1 19 - 20 1292 55
          XM_012957201.2 XP_012812655.1 19 - 20 1291 55
          XM_012957202.1 XP_012812656.1 19 - 20 1291 55
          XM_012957204.1 XP_012812658.1 19 - 20 1292 55
          XM_012957205.1 XP_012812659.1 19 - 20 1291 55
          XM_012957206.1 XP_012812660.1 19 - 20 1291 55
          XM_012957207.1 XP_012812661.1 19 - 20 1292 55
          XM_012957208.1 XP_012812662.1 19 - 20 1292 55
          XM_012957209.1 XP_012812663.1 19 - 20 1292 55
          XM_012957210.2 XP_012812664.1 19 - 20 1292 55
          XM_012957211.1 XP_012812665.1 19 - 20 1292 55
          XM_018091327.1 XP_017946816.1 19 - 20 1292 55
          GenBank mRNA DQ523561.1 - 17 - 18 1104 55

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons

          Panther Classification:

          Molecular Function -

          structural protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline