Search
Search

lman2| Gene Symbol: lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078728.1 | NP_001072196.1 | 7 - 8 | 863 | 61 | |
| XM_018092150.1 | XP_017947639.1 | 8 - 9 | 992 | 61 | ||
| XM_018092151.1 | XP_017947640.1 | 7 - 8 | 771 | 61 | ||
| XM_018092152.1 | XP_017947641.1 | 7 - 8 | 835 | 61 | ||
| GenBank mRNA | BC118738.1 | - | 7 - 8 | 863 | 61 | |
| CU075425.1 | - | 7 - 8 | 849 | 61 | ||
Set Membership: |
|