Search
Search

drc7| Gene Symbol: drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078863.1 | NP_001072331.1 | 16 - 17 | 2432 | 60 | |
| XM_012961306.2 | XP_012816760.1 | 17 - 18 | 3309 | 60 | ||
| XM_012961307.2 | XP_012816761.1 | 16 - 17 | 2673 | 60 | ||
| XM_012961308.2 | XP_012816762.1 | 17 - 18 | 3302 | 60 | ||
| XM_012961311.2 | XP_012816765.1 | 18 - 19 | 3174 | 60 | ||
| XM_012961312.2 | XP_012816766.1 | 17 - 18 | 3080 | 60 | ||
| GenBank mRNA | BC121408.1 | - | 16 - 17 | 2432 | 60 | |
Set Membership: |
|