Search
Search

scrn2| Gene Symbol: scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078981.1 | NP_001072449.1 | 7 - 8 | 1270 | 94 | |
| XM_012952558.2 | XP_012808012.1 | 7 - 8 | 1379 | 94 | ||
| GenBank mRNA | BC121706.1 | - | 7 - 8 | 1270 | 94 | |
Set Membership: |
|