Search
Search

adnp2| Gene Symbol: adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079010.1 | NP_001072478.1 | 2 - 3 | 374 | 98 | |
| GenBank mRNA | BC121900.1 | - | 2 - 3 | 374 | 98 | |
| CR848432.2 | - | 2 - 3 | 350 | 98 | ||
Set Membership: |
|