Search
Search

ptbp2| Gene Symbol: ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079011.1 | NP_001072479.1 | 8 - 9 | 989 | 74 | |
| XM_012961324.2 | XP_012816778.1 | 8 - 9 | 1057 | 74 | ||
| XM_018093092.1 | XP_017948581.1 | 7 - 8 | 980 | 74 | ||
| GenBank mRNA | BC121903.1 | - | 8 - 9 | 989 | 74 | |
| CT027943.1 | - | 8 - 9 | 981 | 74 | ||
Set Membership: |
|