Search
Search

bcl11a| Gene Symbol: bcl11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079189.1 | NP_001072657.1 | 1 - 2 | 183 | 61 | |
| XM_012963166.2 | XP_012818620.1 | 1 - 2 | 733 | 61 | ||
| XM_018093948.1 | XP_017949437.1 | 1 - 2 | 733 | 61 | ||
| XM_018093949.1 | XP_017949438.1 | 1 - 2 | 734 | 61 | ||
| GenBank mRNA | BC123944.1 | - | 1 - 2 | 183 | 61 | |
Set Membership: |
|