Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Xt03718834_g1
          See other CAMK2B GE Assays ›
          Gene Symbol:
          camk2b
          Gene Name:
          calcium/calmodulin dependent protein kinase (CaM kinase) II beta
          Gene Aliases:
          cam2, camk2, camk2g, camkII, camkb
          Chromosome Location:
          Chr.3: 108079183 - 108160510 on Build UCB_Xtro_10.0
          Species:
          Xenopus tropicalis
          Species Specific ID (Flybase ID):
          -
          Assay ID Xt03718834_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: camk2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079449.1 NP_001072917.1 11 - 12 1290 92
          XM_012959302.2 XP_012814756.1 11 - 12 1306 92
          XM_012959303.1 XP_012814757.1 11 - 12 1307 92
          XM_012959304.2 XP_012814758.1 11 - 12 1306 92
          XM_012959305.2 XP_012814759.1 11 - 12 1306 92
          XM_012959306.2 XP_012814760.1 11 - 12 1306 92
          XM_012959308.1 XP_012814762.1 11 - 12 1306 92
          XM_012959309.2 XP_012814763.1 11 - 12 1306 92
          XM_012959310.1 XP_012814764.1 11 - 12 1306 92
          XM_012959311.1 XP_012814765.1 11 - 12 1306 92
          XM_012959312.1 XP_012814766.1 11 - 12 1307 92
          XM_012959313.2 XP_012814767.1 11 - 12 1306 92
          XM_012959314.2 XP_012814768.1 11 - 12 1306 92
          XM_012959315.2 XP_012814769.1 11 - 12 1306 92
          XM_012959316.1 XP_012814770.1 11 - 12 1307 92
          XM_012959317.1 XP_012814771.1 11 - 12 1307 92
          XM_012959318.2 XP_012814772.1 11 - 12 1306 92
          XM_012959319.2 XP_012814773.1 11 - 12 1306 92
          XM_012959320.2 XP_012814774.1 11 - 12 1306 92
          XM_012959321.2 XP_012814775.1 11 - 12 1306 92
          XM_012959322.1 XP_012814776.1 11 - 12 1306 92
          XM_012959323.1 XP_012814777.1 11 - 12 1307 92
          GenBank mRNA BC121362.1 - 11 - 12 1290 92
          BC136217.1 - 11 - 12 1271 92

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline