Search
Search

becn1| Gene Symbol: becn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033940.2 | NP_001029112.1 | 5 - 6 | 462 | 74 | |
| XM_018097768.1 | XP_017953257.1 | 5 - 6 | 462 | 74 | ||
| GenBank mRNA | AM051356.1 | - | 5 - 6 | 468 | 74 | |
| BC135115.1 | - | 5 - 6 | 462 | 74 | ||
| CR760359.2 | - | 5 - 6 | 442 | 74 | ||
Set Membership: |
|