Search
Search

tomm22| Gene Symbol: tomm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045787.1 | NP_001039252.1 | 1 - 2 | 222 | 65 | |
| GenBank mRNA | BC160473.1 | - | 1 - 2 | 173 | 65 | |
| CR942798.2 | - | 1 - 2 | 222 | 65 | ||
| CT025331.2 | - | 1 - 2 | 83 | 65 | ||
Set Membership: |
|