Search
Search

arhgef12| Gene Symbol: arhgef12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079028.1 | NP_001072496.1 | 19 - 20 | 1861 | 58 | |
| GenBank mRNA | BC121927.1 | - | 19 - 20 | 1861 | 58 | |
Set Membership: |
|