Search
Search

sesn2| Gene Symbol: sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079562.1 | NP_001073030.1 | 2 - 3 | 285 | 68 | |
| XM_012957122.2 | XP_012812576.2 | 3 - 4 | 505 | 68 | ||
| XM_018091311.1 | XP_017946800.1 | 3 - 4 | 505 | 68 | ||
| XM_018091312.1 | XP_017946801.1 | 2 - 3 | 334 | 68 | ||
| GenBank mRNA | BC121329.1 | - | 2 - 3 | 285 | 68 | |
| CR760547.2 | - | 2 - 3 | 208 | 68 | ||
Set Membership: |
|