Search
Search

bmt2| Gene Symbol: bmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083350.2 | NP_001076819.1 | 1 - 2 | 228 | 62 | |
| GenBank mRNA | BC088770.1 | - | 1 - 2 | 167 | 62 | |
| BC135938.1 | - | 1 - 2 | 180 | 62 | ||
| DN093113.1 | - | 1 - 2 | 228 | 62 | ||
Set Membership: |
|