Search
Search

armc9| Gene Symbol: armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097173.1 | NP_001090642.1 | 4 - 5 | 536 | 65 | |
| XM_012962522.2 | XP_012817976.1 | 5 - 6 | 673 | 65 | ||
| XM_012962524.2 | XP_012817978.1 | 5 - 6 | 618 | 65 | ||
| XM_012962525.2 | XP_012817979.1 | 5 - 6 | 669 | 65 | ||
| XM_012962526.2 | XP_012817980.1 | 5 - 6 | 670 | 65 | ||
| XM_018093720.1 | XP_017949209.1 | 6 - 7 | 748 | 65 | ||
| GenBank mRNA | BC125807.1 | - | 4 - 5 | 536 | 65 | |
| CT025220.2 | - | 5 - 6 | 600 | 65 | ||
Set Membership: |
|