Search
Search

btbd3| Gene Symbol: btbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097228.1 | NP_001090697.1 | 1 - 2 | 627 | 80 | |
| XM_012962527.2 | XP_012817981.1 | 1 - 2 | 763 | 80 | ||
| XM_012962528.2 | XP_012817982.1 | 2 - 3 | 590 | 80 | ||
| GenBank mRNA | BC128618.1 | - | 1 - 2 | 627 | 80 | |
| CU025012.1 | - | 1 - 2 | 656 | 80 | ||
Set Membership: |
|