Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097314.1 | NP_001090783.1 | 11 - 12 | 1634 | 87 | |
XM_012964140.2 | XP_012819594.1 | 11 - 12 | 1391 | 87 | ||
XM_012964141.2 | XP_012819595.1 | 12 - 13 | 1448 | 87 | ||
GenBank mRNA | BC135164.1 | - | 11 - 12 | 1634 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |