Search
Search

cdk16| Gene Symbol: cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097316.1 | NP_001090785.1 | 13 - 14 | 1399 | 86 | |
| GenBank mRNA | BC135168.1 | - | 13 - 14 | 1399 | 86 | |
Set Membership: |
|