Search
Search

ncam1| Gene Symbol: ncam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018095957.1 | XP_017951446.1 | 5 - 6 | 2162 | 80 | |
| XM_018095958.1 | XP_017951447.1 | 5 - 6 | 2082 | 80 | ||
| XM_018095959.1 | XP_017951448.1 | 5 - 6 | 2160 | 80 | ||
| XM_018095960.1 | XP_017951449.1 | 5 - 6 | 2161 | 80 | ||
| XM_018095961.1 | XP_017951450.1 | 5 - 6 | 2078 | 80 | ||
| XM_018095962.1 | XP_017951451.1 | 5 - 6 | 2159 | 80 | ||
| GenBank mRNA | BC135314.1 | - | 5 - 6 | 852 | 80 | |
Set Membership: |
|