Search
Search

angptl2| Gene Symbol: angptl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097403.1 | NP_001090872.1 | 3 - 4 | 1372 | 68 | |
| XM_012969751.2 | XP_012825205.1 | 3 - 4 | 1162 | 68 | ||
| GenBank mRNA | BC135324.1 | - | 3 - 4 | 1372 | 68 | |
Set Membership: |
|