Search
Search

snx15| Gene Symbol: snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100197.1 | NP_001093667.1 | 7 - 8 | 941 | 89 | |
| GenBank mRNA | BC135952.1 | - | 7 - 8 | 941 | 89 | |
| CR942341.2 | - | 7 - 8 | 904 | 89 | ||
Set Membership: |
|