Search
Search

otub2| Gene Symbol: otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101812.1 | NP_001095282.1 | 3 - 4 | 233 | 76 | |
| XM_012967849.2 | XP_012823303.2 | 10 - 11 | 3397 | 76 | ||
| GenBank mRNA | BC135508.1 | - | 3 - 4 | 233 | 76 | |
Set Membership: |
|