Search
Search

fli1| Gene Symbol: fli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102719.1 | NP_001096189.1 | 3 - 4 | 610 | 83 | |
| XM_012965868.2 | XP_012821322.1 | 3 - 4 | 741 | 83 | ||
| XM_012965869.2 | XP_012821323.1 | 3 - 4 | 598 | 83 | ||
| XM_018095217.1 | XP_017950706.1 | 3 - 4 | 800 | 83 | ||
| GenBank mRNA | BC135689.1 | - | 3 - 4 | 610 | 83 | |
Set Membership: |
|