Search
Search

olfm2| Gene Symbol: olfm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102802.1 | NP_001096272.1 | 5 - 6 | 740 | 61 | |
| XM_012953000.1 | XP_012808454.1 | 5 - 6 | 963 | 61 | ||
| GenBank mRNA | BC135503.1 | - | 5 - 6 | 740 | 61 | |
Set Membership: |
|