Search
Search

tmem245| Gene Symbol: tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012953820.2 | XP_012809274.1 | 9 - 10 | 2961 | 76 | |
| XM_018089411.1 | XP_017944900.1 | 8 - 9 | 2936 | 76 | ||
| GenBank mRNA | BC135929.1 | - | 8 - 9 | 1574 | 76 | |
Set Membership: |
|