Search
Search

atg13| Gene Symbol: atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102950.1 | NP_001096420.1 | 5 - 6 | 563 | 66 | |
| XM_012960476.2 | XP_012815930.1 | 5 - 6 | 676 | 66 | ||
| XM_012960477.2 | XP_012815931.1 | 5 - 6 | 676 | 66 | ||
| XM_012960478.2 | XP_012815932.1 | 5 - 6 | 675 | 66 | ||
| GenBank mRNA | BC136085.1 | - | 5 - 6 | 563 | 66 | |
| CR942657.1 | - | 3 - 4 | 286 | 66 | ||
Set Membership: |
|