Search
Search

slc25a18| Gene Symbol: slc25a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102958.1 | NP_001096428.1 | 9 - 10 | 1026 | 89 | |
| XM_012958457.2 | XP_012813911.1 | 10 - 11 | 1103 | 89 | ||
| XM_012958458.2 | XP_012813912.1 | 10 - 11 | 1093 | 89 | ||
| XM_012958459.2 | XP_012813913.1 | 9 - 10 | 1050 | 89 | ||
| XM_012958460.2 | XP_012813914.1 | 8 - 9 | 801 | 89 | ||
| XM_018089438.1 | XP_017944927.1 | 7 - 8 | 816 | 89 | ||
| GenBank mRNA | BC136106.1 | - | 9 - 10 | 1026 | 89 | |
Set Membership: |
|