Search
Search

tmem82| Gene Symbol: tmem82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016278.3 | NP_001016278.2 | 236 | 75 | ||
| GenBank mRNA | BC158423.1 | - | 236 | 75 | ||
| CR761613.2 | - | 230 | 75 | |||
Set Membership: |
|