Search
Search

c1orf50| Gene Symbol: c1orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016665.3 | NP_001016665.1 | 2 - 3 | 197 | 77 | |
| XM_012966617.2 | XP_012822071.1 | 3 - 4 | 381 | 77 | ||
| GenBank mRNA | BC158946.1 | - | 2 - 3 | 197 | 77 | |
| BC171198.1 | - | 2 - 3 | 188 | 77 | ||
| BC171202.1 | - | 2 - 3 | 188 | 77 | ||
| CR760289.2 | - | 3 - 4 | 357 | 77 | ||
Set Membership: |
|