Search
Search

zic2| Gene Symbol: zic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017025.3 | NP_001017025.2 | 2 - 3 | 1566 | 76 | |
| GenBank mRNA | CR762065.2 | - | 2 - 3 | 401 | 76 | |
| CT025301.2 | - | 2 - 3 | 1566 | 76 | ||
Set Membership: |
|