Search
Search

ankrd22| Gene Symbol: ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113090.1 | NP_001106561.1 | 3 - 4 | 391 | 100 | |
| GenBank mRNA | BC154948.1 | - | 3 - 4 | 391 | 100 | |
Set Membership: |
|