Search
Search

rilp| Gene Symbol: rilp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113125.1 | NP_001106596.1 | 7 - 8 | 1111 | 63 | |
| XM_012956373.2 | XP_012811827.1 | 7 - 8 | 1179 | 63 | ||
| GenBank mRNA | BC155426.1 | - | 7 - 8 | 1111 | 63 | |
Set Membership: |
|