Search
Search

c4a| Gene Symbol: c4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113684.1 | NP_001107156.1 | 31 - 32 | 4028 | 89 | |
| XM_012967939.2 | XP_012823393.1 | 32 - 33 | 4070 | 89 | ||
| GenBank mRNA | BC155689.1 | - | 31 - 32 | 4028 | 89 | |
Set Membership: |
|