Search
Search

abcc3| Gene Symbol: abcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113829.1 | NP_001107301.1 | 10 - 11 | 1631 | 65 | |
| XM_012971517.2 | XP_012826971.1 | 11 - 12 | 2153 | 65 | ||
| XM_012971518.2 | XP_012826972.1 | 10 - 11 | 2339 | 65 | ||
| GenBank mRNA | BC122987.1 | - | 9 - 10 | 1443 | 65 | |
| BC155696.1 | - | 10 - 11 | 1631 | 65 | ||
Set Membership: |
|