Search
Search

phyhd1| Gene Symbol: phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114066.1 | NP_001107538.1 | 10 - 11 | 843 | 65 | |
| GenBank mRNA | BC157790.1 | - | 10 - 11 | 843 | 65 | |
| BC158303.1 | - | 3 - 4 | 750 | 65 | ||
| CU075892.1 | - | 10 - 11 | 841 | 65 | ||
Set Membership: |
|