Search
Search

cmc1| Gene Symbol: cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114070.1 | NP_001107542.1 | 3 - 4 | 277 | 87 | |
| GenBank mRNA | BC157800.1 | - | 3 - 4 | 277 | 87 | |
| BC171026.1 | - | 3 - 4 | 268 | 87 | ||
| BC171080.1 | - | 3 - 4 | 268 | 87 | ||
| CU075862.1 | - | 3 - 4 | 238 | 87 | ||
Set Membership: |
|